Anti-IL6 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX10441
89286-756EA 620.33 USD
89286-756
Anti-IL6 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to ARTS

WB: Use at a dilution of 1/15000. Detects a band of approximately 32 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: IL6
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR