Anti-IL6 Rabbit Polyclonal Antibody

Supplier: Genetex

GTX104407
89319-170EA 394.79 USD
89319-170
Anti-IL6 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to GRP75 (heat shock 70kDa protein 9 (mortalin))

Suggested starting dilutions are as follows: WB: 1:500-1:3000, IP: 1:100-1:500. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: IL6
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR