Anti-IVD Mouse Monoclonal Antibody [clone: 3B9]

Supplier: GeneTex
GTX10439
89336-826EA 358 USD
89336-826
Anti-IVD Mouse Monoclonal Antibody [clone: 3B9]
Mouse monoclonal [3B9] to 14-3-3 theta/tau

For IP: Use at an assay dependent dilution. For WB: Use at a dilution of 1:5,000. Detects a band of approximately 31 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: IVD
Clonality: Monoclonal
Clone: 3B9
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Dog, Monkey, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR