Anti-KDM4C Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX104380
89319-164EA 582.76 USD
89319-164
Anti-KDM4C Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to RAB3B (RAB3B, member RAS oncogene family)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: KDM4C
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR