Anti-KHK Rabbit Polyclonal Antibody

GTX10434
89317-184EA 386.18 USD
89317-184
Anti-KHK Rabbit Polyclonal Antibody
Rabbit polyclonal to DNase I

WB: Use at a concentration of 1 ?g/ml. Detects a band of approximately 33 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: KHK
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR