Anti-LEMD3 Rabbit Polyclonal Antibody

Supplier: Genetex

GTX104120
89316-422EA 395.36 USD
89316-422
Anti-LEMD3 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to DHDH (dihydrodiol dehydrogenase (dimeric))

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: LEMD3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Dog, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR