Anti-LEMD3 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX104118
89319-122EA 582.76 USD
89319-122
Anti-LEMD3 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to IL3RB (colony stimulating factor 2 receptor, beta, low-affinity (granulocyte-macrophage))

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: LEMD3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Dog, Monkey, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR