Anti-LOX Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX103976
89319-086EA 427.1 USD
89319-086
Anti-LOX Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to Prostasin (protease, serine, 8)

Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: LOX
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR