Anti-LTA4H Rat Monoclonal Antibody [clone: 53405.111]

Supplier: Genetex

GTX10394
89315-990EA 357.84 USD
89315-990
Anti-LTA4H Rat Monoclonal Antibody [clone: 53405.111]
Antibodies
Rat monoclonal [53405.111] to RANTES

Recommended Starting Dilutions:

For Inhibition assay: Use recombinant mouse RANTES at a concentration of 0.05 ?g/ml and antibody at concentrations from 0.001 to 10 ?g/ml.

For ICC/IF: Use at a concentration of 8-25 ?g/ml.

Optimal titers for applications should be determined by the researcher.

Type: Primary
Antigen: LTA4H
Clonality: Monoclonal
Clone: 53405.111
Conjugation:
Epitope:
Host: Rat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR