Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-LTA4H Mouse Monoclonal Antibody [clone: 24822.11]

Supplier: Genetex

GTX10390
89315-988EA 325.3 USD InStock
89315-988
Anti-LTA4H Mouse Monoclonal Antibody [clone: 24822.11]
Antibodies
Mouse monoclonal [24822.11] to MCP1

Neut: Use at a concentration of 0.1 μg/ml. Human MCP1 is tested for its ability to neutralize the chemoattractant activity of rhMCP1 for human monocytes. The ND50 of the antibody is defined as the concentration of antibody resulting in a one-half maximal inhibition of bioactivity of rhMCP1 that was present at a concentration just high enough to elicit a maximum response. In this bioassay, rhMCP1 was preincubated with various dilutions of the antibody for 1 hour at 37 °C in a 96-well microtiter plate. Then, the MCP1 antibody solution containing rhMCP1 at a final concentration of 0.1 μg/ml was transferred to the lower compartment of a 96-well chemotaxis chamber. The chemotaxis chamber was assembled using a PVP-free polycarbonate filter and the monocytes were added to the top chamber. The chamber was incubated for 90 minutes at 37 °C in a 5% CO2 humidified incubator. The chamber was disassembled and the filter fixed and stained. The optical density of the filter, which is proportional to the number of cells that migrated across the filter, was read with a microtiter plate reader set at 540 nm. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: LTA4H
Clonality: Monoclonal
Clone: 24822.11
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty