Anti-LTA4H Rabbit Polyclonal Antibody

Supplier: Genetex

GTX103880
89319-074EA 394.91 USD
89319-074
Anti-LTA4H Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to ZNF157 (zinc finger protein 157)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: LTA4H
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Dog, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR