Anti-MAP2K4 Goat Polyclonal Antibody

Supplier: GeneTex
GTX10384
89336-824EA 659.34 USD
89336-824
Anti-MAP2K4 Goat Polyclonal Antibody
Antibodies
Goat polyclonal to Macrophage Inflammatory Protein 1 beta

ELISA: Use at a concentration of 0.125 ?g/ml to detect 1ng/well of recombinant human MIP1 beta. Neut: Use at a concentration of 30 ?g/ml. WB: Use at a concentration of 1 ?g/ml to detect 2ng/lane of recombinant human MIP1 beta under non-reducing conditions. Predicted molecular weight: 11 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: MAP2K4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR