Anti-MAPK1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX103832
89319-066EA 642.92 USD
89319-066
Anti-MAPK1 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to ZNF7 (zinc finger protein 7)

Suggested starting dilutions are as follows: WB: 1:500-1:3000, ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: MAPK1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Dog, Monkey, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR