Anti-MAPK1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX103808
89319-064EA 582.76 USD
89319-064
Anti-MAPK1 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to ENO3 (enolase 3 (beta, muscle))

Suggested starting dilutions are as follows: WB: 1:5000-1:20000, ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: MAPK1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Dog, Monkey, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR