Anti-MCL1 Goat Polyclonal Antibody

Supplier: GeneTex
GTX10374
89285-966EA 362.05 USD
89285-966
Anti-MCL1 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: MCL1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR