Anti-MCL1 Rabbit Polyclonal Antibody

GTX103738
89308-654EA 176.03 USD
89308-654
Anti-MCL1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to FUT9 (fucosyltransferase 9 (alpha (1,3) fucosyltransferase))

Antibody reactive against recombinant protein, detects immunogen. Further validation in progress.

Type: Primary
Antigen: MCL1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR