Anti-MICAL1 Goat Polyclonal Antibody

Supplier: GeneTex
GTX10367
89278-774EA 742.61 USD
89278-774
Anti-MICAL1 Goat Polyclonal Antibody
Antibodies
Goat polyclonal to CXCL3

ELISA: Use at a concentration of 0.5 - 1 ?g/ml. The detection limit for rat CINC2 beta is approximately 0.5 ng/well. WB: Use at a concentration of 0.1 - 0.2 ?g/ml. Predicted molecular weight: 7.6 kDa. The detection limit of rat CINC2 beta is approximately 5 ng/lane and 25 ng/lane under non-reducing and reducing conditions, respectively. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: MICAL1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR