Anti-MIOX Rabbit Polyclonal Antibody

Supplier: Genetex

GTX103654
89319-030EA 394.91 USD
89319-030
Anti-MIOX Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to Nectin 3 (poliovirus receptor-related 3)

Suggested starting dilutions are as follows: WB: 1:1000-1:10000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: MIOX
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR