Anti-MDM2 Mouse Monoclonal Antibody [clone: MD-219]

Supplier: Genetex

GTX10344
89316-410EA 358.16 USD
89316-410
Anti-MDM2 Mouse Monoclonal Antibody [clone: MD-219]
Antibodies
Mouse monoclonal [MD-219] to MDM2

For ELISA: Use at an assay dependent dilution. For FACS: 1?g/10^6 cells. For ICC: Use at an assay dependent dilution. For WB: Use at a concentration of 0.5-2 ?g/ml. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: MDM2
Clonality: Monoclonal
Clone: MD-219
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR