Anti-MYC Rabbit Polyclonal Antibody

Supplier: Genetex

GTX103436
89330-406EA 394.91 USD
89330-406
Anti-MYC Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to c-Myc (v-myc myelocytomatosis viral oncogene homolog (avian))

Suggested starting dilutions are as follows: WB: 1:500-1:3000, IP: 1:100-1:500, ICC/IF: 1:100-1:1000, ChIP. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: MYC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR