Anti-RAD23B Rabbit Polyclonal Antibody

Supplier: Genetex

GTX103424
89318-974EA 394.79 USD
89318-974
Anti-RAD23B Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to hHR23b (RAD23 homolog B (S. cerevisiae))

Suggested starting dilutions are as follows: WB: 1:5000-1:20000, IHC-P: 1:100-1:1000, ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: RAD23B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR