Anti-ALDH6A1 Rabbit Polyclonal Antibody

Supplier: Genetex

GTX103405
89318-968EA 394.91 USD
89318-968
Anti-ALDH6A1 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to ALDH6A1 (aldehyde dehydrogenase 6 family, member A1)

Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: ALDH6A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR