Anti-NOG Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX103314
89318-950EA 582.76 USD
89318-950
Anti-NOG Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to SLC4A8 (solute carrier family 4, sodium bicarbonate cotransporter, member 8)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: NOG
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR