Anti-NRBP1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX103228
89318-930EA 582.76 USD
89318-930
Anti-NRBP1 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to MSH3 (mutS homolog 3 (E. coli))

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: NRBP1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR