Anti-NT5DC1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX103181
89318-924EA 394.91 USD
89318-924
Anti-NT5DC1 Rabbit Polyclonal Antibody
Rabbit Polyclonal antibody to DNA polymerase beta (polymerase (DNA directed), beta)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: NT5DC1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Dog, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR