Anti-NTRK2 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX103126
89308-532EA 176.03 USD
89308-532
Anti-NTRK2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to ILT-4 (leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), member 2)

Antibody reactive against recombinant protein, detects immunogen. Further validation in progress.

Type: Primary
Antigen: NTRK2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Dog, Monkey, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR