Anti-NXNL2 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX103071
89318-908EA 448.72 USD
89318-908
Anti-NXNL2 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to MCM2 (minichromosome maintenance complex component 2)

Suggested starting dilutions are as follows: WB: 1:500-1:3000, IP: 1:100-1:500. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: NXNL2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR