Anti-ODC1 Rabbit Polyclonal Antibody

GTX103070
89308-520EA 176.03 USD
89308-520
Anti-ODC1 Rabbit Polyclonal Antibody
Rabbit Polyclonal antibody to TFIIH p34 (general transcription factor IIH, polypeptide 3, 34kDa)

Antibody reactive against recombinant protein, detects immunogen. Further validation in progress.

Type: Primary
Antigen: ODC1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR