Anti-OSBPL11 Rabbit Polyclonal Antibody

Supplier: Genetex

GTX10306
89329-656EA 395.41 USD
89329-656
Anti-OSBPL11 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: OSBPL11
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR