Anti-OSBPL11 Rabbit Polyclonal Antibody

GTX103051
89308-514EA 176.03 USD
89308-514
Anti-OSBPL11 Rabbit Polyclonal Antibody
Rabbit polyclonal antibody to Thrombospondin 5 (cartilage oligomeric matrix protein)

Antibody reactive against recombinant protein, detects immunogen. Further validation in progress.

Type: Primary
Antigen: OSBPL11
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR