Anti-OSBPL11 Rabbit Polyclonal Antibody

GTX10304
89291-564EA 399.48 USD
89291-564
Anti-OSBPL11 Rabbit Polyclonal Antibody
Rabbit polyclonal to metabotropic Glutamate Receptor 2

WB: Use at a dilution of 1/1000. Predicted molecular weight: 104.6 kDa. GTX10304 has been only shown to work by Western blot in highly expressing transfected cells using standard protocols. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: OSBPL11
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR