Anti-OXSR1 Rabbit Polyclonal Antibody

Supplier: Genetex

GTX103011
89318-898EA 394.91 USD
89318-898
Anti-OXSR1 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to HSP47 (serpin peptidase inhibitor, clade H (heat shock protein 47), member 1, (collagen binding protein 1))

Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: OXSR1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Dog, Monkey, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR