Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PANK2 Mouse Monoclonal Antibody [clone: 6A15]

Supplier: Genetex

GTX10299
89300-300EA 367.83 USD InStock
89300-300
Anti-PANK2 Mouse Monoclonal Antibody [clone: 6A15]
Antibodies
Mouse Monoclonal antibody [6A15] to FMR1 (Drosophila)

Optimal dilutions should be determined experimentally by the researcher.

Type: Primary
Antigen: PANK2
Clonality: Monoclonal
Clone: 6A15
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty