Anti-PCSK7 Rabbit Polyclonal Antibody

Supplier: Genetex

GTX102957
89318-886EA 394.91 USD
89318-886
Anti-PCSK7 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to NCF1 (neutrophil cytosolic factor 1)

Suggested starting dilutions are as follows: WB: 1:1000-1:10000, IHC-P: 1:100-1:1000, ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: PCSK7
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR