Anti-PIK3AP1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX102825
89316-398EA 727.73 USD
89316-398
Anti-PIK3AP1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to PSCD1 (pleckstrin homology, Sec7 and coiled-coil domains 1)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: PIK3AP1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Monkey, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR