Anti-PKLR Rabbit Polyclonal Antibody

Supplier: Genetex

GTX10277
89337-276EA 394.88 USD
89337-276
Anti-PKLR Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to Nociceptin

IHC-Fr: Use at a dilution of 1/500. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: PKLR
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR