Anti-POR Goat Polyclonal Antibody

Supplier: GeneTex
GTX10268
89287-850EA 607.87 USD
89287-850
Anti-POR Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to TIP30 / CC3

Approx 28kDa band observed in lysates of cell line HeLa (calculated MW of 27.0kDa according to NP_006401.3). Recommended concentration: 0.3-1µg/ml. This antibody is expected to recognise all three reported isoforms. Variants (NP_006401.3; NP_001091991.1; NP_001091992.1) encode the same isoform.

Type: Primary
Antigen: POR
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR