Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PPP6C Goat Polyclonal Antibody

Supplier: Genetex

GTX10265
89287-848EA 388.49 USD InStock
89287-848
Anti-PPP6C Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to PITPN / PITP alpha

Approx 32kDa band observed in 293 and Human Liver lysates (predicted MW of 31kDa according to NP_006215). Recommended for use at 1-3µg/ml.

Type: Primary
Antigen: PPP6C
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty