Anti-PRKAR1A Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX102623
89318-790EA 427.1 USD
89318-790
Anti-PRKAR1A Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to DYRK1B (dual-specificity tyrosine-(Y)-phosphorylation regulated kinase 1B)

Suggested starting dilutions are as follows: WB: 1:500-1:3000, ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: PRKAR1A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR