Anti-PRKG1 Goat Polyclonal Antibody

Supplier: Genetex

GTX10256
89287-844EA 427.55 USD
89287-844
Anti-PRKG1 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to SSX2IP

We find no specific signal but low background (at antibody concentration up to 1µg/ml) in lysates of cell line K562.

Type: Primary
Antigen: PRKG1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR