Anti-PRKY Mouse Monoclonal Antibody [clone: 7F8]

GTX10254
89336-528EA 258.81 USD
89336-528
Anti-PRKY Mouse Monoclonal Antibody [clone: 7F8]
Antibodies
Mouse monoclonal antibody [7F8] to CD71

ELISA: Use at an assay dependent dilution. Recommended pair antibody: GTX10254 (coating) with GTX10251 (detection). GTX10254 (detection) with GTX10249 (coating). Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: PRKY
Clonality: Monoclonal
Clone: 7F8
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR