Anti-PRKY Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX102539
89318-780EA 582.76 USD
89318-780
Anti-PRKY Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to PKC gamma (protein kinase C, gamma)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: PRKY
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR