Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PRKY Mouse Monoclonal Antibody [clone: 5B11]

Supplier: Genetex
GTX10253
89336-526
Anti-PRKY Mouse Monoclonal Antibody [clone: 5B11]
Mouse monoclonal antibody [5B11] to CD71

ELISA: Use at an assay dependent dilution. Recommended pair antibody: GTX10253 (coating) with GTX10252 (detection). Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: PRKY
Clonality: Monoclonal
Clone: 5B11
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human

Order Now


 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty