Anti-PRL Mouse Monoclonal Antibody [clone: 2B6]

Supplier: GeneTex
GTX10252
89336-524EA 759 USD
89336-524
Anti-PRL Mouse Monoclonal Antibody [clone: 2B6]
Antibodies
Mouse monoclonal antibody [2B6] to CD71

ELISA: Use at an assay dependent dilution. Recommended pair antibody: GTX10252 (coating) with GTX10249 (detection).ab 10253 (detection) with GTX10252 (coating). Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: PRL
Clonality: Monoclonal
Clone: 2B6
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR