Anti-PROM2 Rabbit Polyclonal Antibody

GTX102517
89308-368EA 176.03 USD
89308-368
Anti-PROM2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to GCS-alpha-3 (guanylate cyclase 1, soluble, alpha 3)

Antibody reactive against recombinant protein, detects immunogen. Further validation in progress.

Type: Primary
Antigen: PROM2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR