Anti-PROM2 Mouse Monoclonal Antibody [clone: 23D10]

Supplier: GeneTex

GTX10251
89336-522EA 293.4 USD
89336-522
Anti-PROM2 Mouse Monoclonal Antibody [clone: 23D10]
Antibodies
Mouse monoclonal antibody [23D10] to CD71

ELISA: Use at an assay dependent dilution. Recommended pair antibody: GTX10251 (detection) with GTX10254 (coating). Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: PROM2
Clonality: Monoclonal
Clone: 23D10
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR