Anti-PRPSAP2 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX102500
89318-770EA 582.76 USD
89318-770
Anti-PRPSAP2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to Calpain 3 (calpain 3, (p94))

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: PRPSAP2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR