Anti-PRTG Mouse Monoclonal Antibody [clone: 130000]

Supplier: GeneTex
GTX10250
89336-520EA 759 USD
89336-520
Anti-PRTG Mouse Monoclonal Antibody [clone: 130000]
Antibodies
Mouse monoclonal antibody [13E4] to CD71

ELISA: Use at an assay dependent dilution. Recommended pair antibody: GTX10250 (detection) with GTX10247 (coating). Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: PRTG
Clonality: Monoclonal
Clone: 130000
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR