Anti-PTCH1 Rabbit Polyclonal Antibody

Supplier: Genetex

GTX102475
89318-760EA 394.91 USD
89318-760
Anti-PTCH1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to NME5 (non-metastatic cells 5, protein expressed in (nucleoside-diphosphate kinase))

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: PTCH1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR