Anti-PTPRE Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX102466
89308-338EA 176.03 USD
89308-338
Anti-PTPRE Rabbit Polyclonal Antibody
Rabbit polyclonal antibody to TSG-6 (tumor necrosis factor, alpha-induced protein 6)

Antibody reactive against recombinant protein, detects immunogen. Further validation in progress.

Type: Primary
Antigen: PTPRE
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR